Learn More
Cayman Chemical Oleic Acid ethyl ester
SDP
Catalog No. 502889562
Encompass_Preferred
Oleic acid ethyl ester is a neutral, more lipid-soluble form of oleic acid.
Catalog No. 50-288-9562 Supplier Cayman Chemical Supplier No. 1000820150 MG
Oleic acid ethyl ester is a neutral, more lipid-soluble form of oleic acid.