Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleic Acid, Food Grade, Spectrum™ Chemical
SDP

Catalog No. 18604879 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-879 4 L Amber Glass Bottle
18-604-881 20 L Amber Glass Bottle
18-604-880 200 L Amber Glass Bottle
18-604-878 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-879 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. O15004LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

O1500, 112-80-1, C18H34O2

Spectrum™ Chemical Oleic Acid, Food Grade

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
IUPAC Name (9E)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

CAS Max % 1
Boiling Point 286.1°C
Packaging Amber Glass Bottle
Quantity 4 L
Saponification Value 196-204
Grade Food Grade
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.