Learn More
Cayman Chemical OleIc AcId methyl ester 500mg
SDP
Catalog No. 502984741
Encompass_Preferred
An esterified version of the free acid which is less water soluble but more amenable for the formulation of oleate-containing diets and dietary supplements
Catalog No. 50-298-4741 Supplier Cayman Chemical Supplier No. 20604500 MG
An esterified version of the free acid which is less water soluble but more amenable for the formulation of oleate-containing diets and dietary supplements