Learn More
Medchemexpress LLC Oleic acid sodium | 143-19-1 | 304.44 | 10 G
SDP
Catalog No. 5000414551
Encompass_Preferred
Sodium oleate (Oleic acid sodium) is an abundant monounsaturated fatty acid sodium and a Na+/K+ ATPase activator. It is suitable for various research applications.
  • For research use only
  • Keep dry and cool
  • Store at room temperature
  • Avoid prolonged storage in solvent at higher temperatures
Catalog No. 50-004-14551 Supplier Medchemexpress LLC Supplier No. HYN1446B10G
Sodium oleate (Oleic acid sodium) is an abundant monounsaturated fatty acid sodium and a Na+/K+ ATPase activator. It is suitable for various research applications.
  • For research use only
  • Keep dry and cool
  • Store at room temperature
  • Avoid prolonged storage in solvent at higher temperatures