Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleic acid sodium | 143-19-1 | 304.44 | 50 G
SDP

Catalog No. 5000414674
Encompass_Preferred

Sodium oleate (Oleic acid sodium) is an abundant monounsaturated fatty acid sodium. It is also known as a Na+/K+ ATPase activator.

  • Activates Na+/K+ ATPase.
  • Promotes cell proliferation and migration in high metastatic cancer cells via enhancing β-oxidation mediated by AMPK activation.
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells.
  • Purity: 99.99%.
  • Appearance: Solid.
  • Color: White to off-white.
  • Formula: C18H33NaO2.
  • Structure classification: Ketones, Aldehydes, Acids.
  • Initial source: Endogenous metabolite.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Store at room temperature, keep dry and cool. In solvent: -80°C 1 year, -20°C 6 months.

Catalog No. 50-004-14674 Supplier Medchemexpress LLC Supplier No. HYN1446B50G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium oleate (Oleic acid sodium) is an abundant monounsaturated fatty acid sodium. It is also known as a Na+/K+ ATPase activator.

  • Activates Na+/K+ ATPase.
  • Promotes cell proliferation and migration in high metastatic cancer cells via enhancing β-oxidation mediated by AMPK activation.
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells.
  • Purity: 99.99%.
  • Appearance: Solid.
  • Color: White to off-white.
  • Formula: C18H33NaO2.
  • Structure classification: Ketones, Aldehydes, Acids.
  • Initial source: Endogenous metabolite.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: Store at room temperature, keep dry and cool. In solvent: -80°C 1 year, -20°C 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.