Learn More
Medchemexpress LLC Oleic acid sodium (sodium oleate) | 143-19-1 | 304.44 | 100 G
SDP
Catalog No. 5000414860
Encompass_Preferred
Sodium oleate (Oleic acid sodium) is an abundant monounsaturated fatty acid sodium and a Na+/K+ ATPase activator. Its active component, oleic acid, influences cell proliferation and migration, showing diverse effects across various cancer cell types.
  • Abundant monounsaturated fatty acid sodium
  • Na+/K+ ATPase activator
  • Used for research purposes only
  • Oleic acid component influences cell proliferation and migration
  • Can promote cell proliferation in highly metastatic cancer cells
  • Can inhibit cancer cell growth in low metastatic carcinoma cells
Catalog No. 50-004-14860 Supplier Medchemexpress LLC Supplier No. HYN1446B100G
Sodium oleate (Oleic acid sodium) is an abundant monounsaturated fatty acid sodium and a Na+/K+ ATPase activator. Its active component, oleic acid, influences cell proliferation and migration, showing diverse effects across various cancer cell types.
  • Abundant monounsaturated fatty acid sodium
  • Na+/K+ ATPase activator
  • Used for research purposes only
  • Oleic acid component influences cell proliferation and migration
  • Can promote cell proliferation in highly metastatic cancer cells
  • Can inhibit cancer cell growth in low metastatic carcinoma cells