Learn More
Cayman Chemical Oleic Acid2 6diisopropylanili
SDP
Catalog No. 502889882
Encompass_Preferred
A hypocholesterolemic ACAT inhibitor (IC50 = 7 nM); dietary administration (0.05%) decreases LDLs and elevates HDL levels in rats and rabbits fed a high fat, high cholesterol diet
Catalog No. 50-288-9882 Supplier Cayman Chemical Supplier No. 1000678210 MG
A hypocholesterolemic ACAT inhibitor (IC50 = 7 nM); dietary administration (0.05%) decreases LDLs and elevates HDL levels in rats and rabbits fed a high fat, high cholesterol diet