Learn More
Cayman Chemical Oleic Acid2 6diisopropylanili
SDP
Supplier:  Cayman Chemical 1000678250 MG
Encompass_Preferred
A hypocholesterolemic ACAT inhibitor (IC50 = 7 nM); dietary administration (0.05%) decreases LDLs and elevates HDL levels in rats and rabbits fed a high fat, high cholesterol diet
Catalog No. 50-289-1447