Learn More
Cayman Chemical Oleic Acid-2,6-diisopropylanilide, Purity: ≥98%, CAS: 140112-65-8, 5 mg
SDP
Catalog No. 502888773
Encompass_Preferred
A hypocholesterolemic ACAT inhibitor (IC50 = 7 nM); dietary administration (0.05%) decreases LDLs and elevates HDL levels in rats and rabbits fed a high fat, high cholesterol diet
Catalog No. 50-288-8773 Supplier Cayman Chemical Supplier No. 100067825 MG
A hypocholesterolemic ACAT inhibitor (IC50 = 7 nM); dietary administration (0.05%) decreases LDLs and elevates HDL levels in rats and rabbits fed a high fat, high cholesterol diet