Learn More
Cayman Chemical Oleic Acidbiotin
SDP
Catalog No. 502890099
Encompass_Preferred
An affinity probe designed to allow oleic acid to be detected in complexes with protein binding partners such as fatty acid binding proteins
Catalog No. 50-289-0099 Supplier Cayman Chemical Supplier No. 10010624100 UG
An affinity probe designed to allow oleic acid to be detected in complexes with protein binding partners such as fatty acid binding proteins