Learn More
Cayman Chemical OleIc AcIdodIum salt 25g
SDP
Catalog No. 502935771
Encompass_Preferred
A monounsaturated fatty acid; a major component of membrane phospholipids; inhibits collagen-stimulated platelet aggregation at 10 UG/ml; inhibits fMLF-induced neutrophil aggregation and degranulation at 5 µM; induces release of intracellular calcium in human platelets at 60 µM; increases TNF-α, IL-8, IL-6, and IL-1β production, neutrophil accumulation, and apoptotic and necrotic cell death in mouse lung
Catalog No. 50-293-5771 Supplier Cayman Chemical Supplier No. 2465925 G
A monounsaturated fatty acid; a major component of membrane phospholipids; inhibits collagen-stimulated platelet aggregation at 10 UG/ml; inhibits fMLF-induced neutrophil aggregation and degranulation at 5 µM; induces release of intracellular calcium in human platelets at 60 µM; increases TNF-α, IL-8, IL-6, and IL-1β production, neutrophil accumulation, and apoptotic and necrotic cell death in mouse lung