Learn More
Cayman Chemical OleIc nhydrIde 100mg
SDP
Catalog No. 502982097
Encompass_Preferred
A fatty acid anhydride; inhibits sphingosine-induced phosphorylation of p32 in Jurkat T cells from 30-100 μM; has been used in the synthesis of various phospholipids and triglycerides
Catalog No. 50-298-2097 Supplier Cayman Chemical Supplier No. 20825100 MG
A fatty acid anhydride; inhibits sphingosine-induced phosphorylation of p32 in Jurkat T cells from 30-100 μM; has been used in the synthesis of various phospholipids and triglycerides