Learn More
Cayman Chemical Oleic Anhydride, Purity: ≥95%, CAS: 24909-72-6, 50 mg
SDP
Catalog No. 502982093
Encompass_Preferred
A fatty acid anhydride; inhibits sphingosine-induced phosphorylation of p32 in Jurkat T cells from 30-100 μM; has been used in the synthesis of various phospholipids and triglycerides
Catalog No. 50-298-2093 Supplier Cayman Chemical Supplier No. 2082550 MG
A fatty acid anhydride; inhibits sphingosine-induced phosphorylation of p32 in Jurkat T cells from 30-100 μM; has been used in the synthesis of various phospholipids and triglycerides