Learn More
Cayman Chemical OlenolIc AcId Acrylate 10mg
SDP
Catalog No. 502937808
Encompass_Preferred
An agonist of 5-HT1A and a derivative of oleanolic acid; acts as an agonist at the 5-HT1A receptor in mouse hippocampal homogenates in a [35S]GTPγS binding assay (EC50 = 0.79 µM); a selective inhibitor of MAO-A over MAO-B (IC50s = 48.8 µM and >100 µM, respectively); reduces immobility in the forced swim test and increases the number of crossings and time spent at the center in the open field test, indicating anti-depressant-like and anxiolytic activities in mice at 10 mg/kg
Catalog No. 50-293-7808 Supplier Cayman Chemical Supplier No. 2720610 MG
An agonist of 5-HT1A and a derivative of oleanolic acid; acts as an agonist at the 5-HT1A receptor in mouse hippocampal homogenates in a [35S]GTPγS binding assay (EC50 = 0.79 µM); a selective inhibitor of MAO-A over MAO-B (IC50s = 48.8 µM and >100 µM, respectively); reduces immobility in the forced swim test and increases the number of crossings and time spent at the center in the open field test, indicating anti-depressant-like and anxiolytic activities in mice at 10 mg/kg