Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleonuezhenide | 112693-21-7 | 1 MG
SDP

Catalog No. 5000420580
Encompass_Preferred

Oleonuezhenide, isolated from Fructus Ligustri Lucidi, is a white to off-white solid with a purity of 99.27%. It has a molecular weight of 1073.01 and a chemical formula of C48H64O27. This compound exerts neuroprotective effects and is soluble in ethanol at ≥ 50 mg/mL.

  • Exerts neuroprotective effects
  • Isolated from Fructus Ligustri Lucidi
  • High purity: 99.27%
  • Molecular weight: 1073.01
  • Solid appearance, white to off-white color
  • Soluble in ethanol

Catalog No. 50-004-20580 Supplier Medchemexpress LLC Supplier No. HYN31451MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleonuezhenide, isolated from Fructus Ligustri Lucidi, is a white to off-white solid with a purity of 99.27%. It has a molecular weight of 1073.01 and a chemical formula of C48H64O27. This compound exerts neuroprotective effects and is soluble in ethanol at ≥ 50 mg/mL.

  • Exerts neuroprotective effects
  • Isolated from Fructus Ligustri Lucidi
  • High purity: 99.27%
  • Molecular weight: 1073.01
  • Solid appearance, white to off-white color
  • Soluble in ethanol

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.