Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleonuezhenide | 112693-21-7 | 5 MG
SDP

Catalog No. 5000420579
Encompass_Preferred

Oleonuezhenide, isolated from Fructus Ligustri Lucidi, is a compound that exerts neuroprotective effects. It is provided as a white to off-white solid and is intended for research use only.

  • Exhibits neuroprotective effects
  • High purity of 99.27%
  • Molecular formula: C48H64O27
  • Molecular weight: 1073.01
  • Soluble in ethanol (≥ 50 mg/mL)
  • Provided with in vivo dissolution protocols

Catalog No. 50-004-20579 Supplier Medchemexpress LLC Supplier No. HYN31455MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleonuezhenide, isolated from Fructus Ligustri Lucidi, is a compound that exerts neuroprotective effects. It is provided as a white to off-white solid and is intended for research use only.

  • Exhibits neuroprotective effects
  • High purity of 99.27%
  • Molecular formula: C48H64O27
  • Molecular weight: 1073.01
  • Soluble in ethanol (≥ 50 mg/mL)
  • Provided with in vivo dissolution protocols

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.