Learn More
American Radiolabeled Chemicals Inc OLEOYL-1-14C COA 50UC
SDP
Catalog No. 5090103632
Encompass_Preferred
Oleoyl-1-14C Coenzyme A 50uCi in 0.01M Na acetate (pH 5.2):ethanol (1:1) under argon, 0.1 mCi/ml, S.A. 40-60 mCi/mmol, M.W. 1032.1, Shipped in Dry Ice
Catalog No. 50-901-03632 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 052750UC
Oleoyl-1-14C Coenzyme A 50uCi in 0.01M Na acetate (pH 5.2):ethanol (1:1) under argon, 0.1 mCi/ml, S.A. 40-60 mCi/mmol, M.W. 1032.1, Shipped in Dry Ice