Learn More
Cayman Chemical Oleoyl 3-carbacyclic Phosphatidic Acid, Purity: ≥95%, CAS: 779333-58-3, 10 mg
SDP
Catalog No. 502891355
Encompass_Preferred
A cyclic LPA analog that contains the 18:1 fatty acid, oleate, at the sn-1 position; at 25 μM, inhibits the transcellular migration of MM1 cells across mesothelial cell monolayers without affecting proliferation; at 0.1-1.0 μM, significantly inhibits autotaxin and antagonizes LPA1 (IC50 = 836 nM) and LPA3 (IC50 = 440 nM)
Catalog No. 50-289-1355 Supplier Cayman Chemical Supplier No. 1001029910 MG
A cyclic LPA analog that contains the 18:1 fatty acid, oleate, at the sn-1 position; at 25 μM, inhibits the transcellular migration of MM1 cells across mesothelial cell monolayers without affecting proliferation; at 0.1-1.0 μM, significantly inhibits autotaxin and antagonizes LPA1 (IC50 = 836 nM) and LPA3 (IC50 = 440 nM)