Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oleoyl chloride

Catalog No. 367850500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-785-0500ML 500 mL
36-785-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-785-0500ML Supplier Sigma Aldrich Supplier No. 367850500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Oleoyl chloride is a fatty acid derivative.oleoyl chloride may be used:
  • In the chemical modification of the jute fibbers to confer hydrophobicity and resistance to biofibers
  • In the synthesis of oleoyl-s-lipoate, via reaction with dihydrolipoate
  • In the preparation of symmetrical triglyceride, 2-oleoyl distearin
  • In the preparation of ergosteryl oleate

Specifications

CAS 112-77-6
Density 0.91 g/mL (at 25°C (literature))
Boiling Point 193°C (4 mmHg)
Molecular Formula C18H33OCl
Refractive Index n20/D 1.463 (literature)
Linear Formula CH3(CH2)7 CH=CH(CH2)7 COCl
MDL Number MFCD00134332
Quantity 500 mL
Molecular Weight (g/mol) 300.91
Percent Purity ≥89%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.