Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Oleoyl Coenzyme A Dipotassium Salt

Catalog No. MP210090401 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210090401 1 mg
ICN10090405 5 mg
MP210090410 10 mg
02-100-90450 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP210090401 Supplier MP Biomedicals™ Supplier No. 0210090401
Add to Cart
Add to Cart

Oleoyl coenzyme A (Oleoyl-CoA) may be used to study the specificity and kinetics of enzymes such as acyl-coenzyme A:cholesterol acyltransferase 1 (ACAT1) involved in long chain fatty acid cholesterol esterification.

  • Coenzyme A functions as an acyl group carrier, acetyl-CoA

  • Coenzyme A functions as an acyl group carrier, acetyl-CoA
TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage -20°C
Form Powder
Quantity 1 mg
Molecular Weight (g/mol) 1031.986
Product Type Biochemicals
Purity or Quality Grade ≥90%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.