Learn More
Cayman Chemical Oleoyl-D-lysine (sodium salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502941757
Encompass_Preferred
An inhibitor of GlyT2a; selectively inhibits GlyT2a (IC50 = 48.3 nM) over GlyT1b (IC50 = >3,000 nM) in Xenopus oocytes expressing the human transporters; reduces allodynia in a nerve ligation rat model of neuropathic pain at 30 mg/kg, i.p.
Catalog No. 50-294-1757 Supplier Cayman Chemical Supplier No. 2786210 MG
An inhibitor of GlyT2a; selectively inhibits GlyT2a (IC50 = 48.3 nM) over GlyT1b (IC50 = >3,000 nM) in Xenopus oocytes expressing the human transporters; reduces allodynia in a nerve ligation rat model of neuropathic pain at 30 mg/kg, i.p.