Learn More
Cayman Chemical Oleoyl Ethanolamide Phosphate
SDP
Supplier:  Cayman Chemical 228861 MG
Encompass_Preferred
An LPA mimetic, LPA receptor agonist, and phosphate ester of OEA; selectively increases [35S]GTPγS binding to HEK293T cell membranes expressing LPA1 and LPA2 over LPA3 receptors; induces calcium mobilization and inhibits forskolin-induced cAMP accumulation in MDA-MB-231 cells (EC50s = 1.2 and 101 nM, respectively)
Catalog No. 50-299-4231