Learn More
Cayman Chemical Oleoyl Ethanolamide-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 1245477-09-1, 5 mg
SDP
Catalog No. 502889423
Encompass_Preferred
An internal standard for the quantification of OEA by GC- or LC-MS.
Catalog No. 50-288-9423 Supplier Cayman Chemical Supplier No. 100078235 MG
An internal standard for the quantification of OEA by GC- or LC-MS.