Learn More
Cayman Chemical Oleoyl Ethanolamide-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 1245477-09-1, 1 mg
SDP
Catalog No. 502890574
Encompass_Preferred
An internal standard for the quantification of OEA by GC- or LC-MS.
Catalog No. 50-289-0574 Supplier Cayman Chemical Supplier No. 100078231 MG
An internal standard for the quantification of OEA by GC- or LC-MS.