Learn More
Cayman Chemical Oleoyl EthnolamIde 100mg
SDP
Catalog No. 502895052
Encompass_Preferred
An analog of the endocannabinoid AEA found in brain tissue and in chocolate whose biosynthesis is reduced in the intestine of rats following food deprivation; an endogenous, potent agonist for PPARα (EC50 = 120 nM in a transactivation assay); systemic administration suppresses food intake and reduces weight gain in rats (10 mg/kg intraperitoneally) and PPARα wild type mice, but not in PPARα knockout mice
Catalog No. 50-289-5052 Supplier Cayman Chemical Supplier No. 90265100 MG
An analog of the endocannabinoid AEA found in brain tissue and in chocolate whose biosynthesis is reduced in the intestine of rats following food deprivation; an endogenous, potent agonist for PPARα (EC50 = 120 nM in a transactivation assay); systemic administration suppresses food intake and reduces weight gain in rats (10 mg/kg intraperitoneally) and PPARα wild type mice, but not in PPARα knockout mice