Learn More
Cayman Chemical Oleoyl Ethanolamide, Purity: ≥98%, CAS: 111-58-0, 500 mg
SDP
Catalog No. 502892997
Encompass_Preferred
An analog of the endocannabinoid AEA found in brain tissue and in chocolate whose biosynthesis is reduced in the intestine of rats following food deprivation; an endogenous, potent agonist for PPARα (EC50 = 120 nM in a transactivation assay); systemic administration suppresses food intake and reduces weight gain in rats (10 mg/kg intraperitoneally) and PPARα wild type mice, but not in PPARα knockout mice
Catalog No. 50-289-2997 Supplier Cayman Chemical Supplier No. 90265500 MG
An analog of the endocannabinoid AEA found in brain tissue and in chocolate whose biosynthesis is reduced in the intestine of rats following food deprivation; an endogenous, potent agonist for PPARα (EC50 = 120 nM in a transactivation assay); systemic administration suppresses food intake and reduces weight gain in rats (10 mg/kg intraperitoneally) and PPARα wild type mice, but not in PPARα knockout mice