Learn More
Cayman Chemical Oleoyl EthnolamIde-d4 100UG
SDP
Catalog No. 502892404
Encompass_Preferred
An internal standard for the quantification of OEA by GC- or LC-MS
Catalog No. 50-289-2404 Supplier Cayman Chemical Supplier No. 9000552100 UG
An internal standard for the quantification of OEA by GC- or LC-MS