Learn More
Cayman Chemical Oleoyl EthnolamIde-d4 1mg
SDP
Catalog No. 502892598
Encompass_Preferred
An internal standard for the quantification of OEA by GC- or LC-MS
Catalog No. 50-289-2598 Supplier Cayman Chemical Supplier No. 90005521 MG
An internal standard for the quantification of OEA by GC- or LC-MS