Learn More
Cayman Chemical Oleoyl EthnolamIde-d4 500UG
SDP
Catalog No. 502897013
Encompass_Preferred
An internal standard for the quantification of OEA by GC- or LC-MS
Catalog No. 50-289-7013 Supplier Cayman Chemical Supplier No. 9000552500 UG
An internal standard for the quantification of OEA by GC- or LC-MS