Learn More
Cayman Chemical Oleoyl Ethanolamide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 946524-36-3, 5 mg
SDP
Catalog No. 502894630
Encompass_Preferred
An internal standard for the quantification of OEA by GC- or LC-MS
Catalog No. 50-289-4630 Supplier Cayman Chemical Supplier No. 90005525 MG
An internal standard for the quantification of OEA by GC- or LC-MS