Learn More
Cayman Chemical Oleoyl Ethyl Amide, Purity: ≥98%, CAS: 85075-82-7, 5 mg
SDP
Catalog No. 502888778
Encompass_Preferred
A selective FAAH inhibitor (IC50 = 5.25 nM in rat brain homogenates) with potential analgesic and anxiolytic activity; does not inhibit acidic PEAase or bind to CB1 or CB2 receptors
Catalog No. 50-288-8778 Supplier Cayman Chemical Supplier No. 100054595 MG
A selective FAAH inhibitor (IC50 = 5.25 nM in rat brain homogenates) with potential analgesic and anxiolytic activity; does not inhibit acidic PEAase or bind to CB1 or CB2 receptors