Learn More
Apexbio Technology LLC Oleoyl Ethyl Amide(Synonyms: Oleoylethanolamide, Oleic acid ethanolamide, OEA, N-Oleoylethanolamine, N-(2-Hydroxyethyl)oleamide), 5mg (solution), CAS: 85075-82-7.
SDP
Catalog No. 5000061231
Encompass_Preferred
Oleoyl Ethyl Amide (CAS 85075-82-7) abbreviated OEtA is a fatty acid amide hydrolase (FAAH) inhibitor displaying analgesic and anxiolytic properties FAAH a membrane-bound serine hydrolase is responsible in vivo for degrading fatty acid amides (FAAs) signaling lipids involved in pain and anxiety modulation In rat brain homogenates OEtA inhibits FAAH activity potently with an IC50 of 5 25 nM without affinity for CB1 or CB2 cannabinoid receptors or inhibition of acidic PEAase activity Animal studies indicate that chronic subcutaneous administration of OEtA reduces bladder overactivity and improves urodynamic parameters supporting its application in research on pain anxiety and bladder dysfunction
Catalog No. 50-000-61231 Supplier Apexbio Technology LLC Supplier No. C42515
Oleoyl Ethyl Amide (CAS 85075-82-7) abbreviated OEtA is a fatty acid amide hydrolase (FAAH) inhibitor displaying analgesic and anxiolytic properties FAAH a membrane-bound serine hydrolase is responsible in vivo for degrading fatty acid amides (FAAs) signaling lipids involved in pain and anxiety modulation In rat brain homogenates OEtA inhibits FAAH activity potently with an IC50 of 5 25 nM without affinity for CB1 or CB2 cannabinoid receptors or inhibition of acidic PEAase activity Animal studies indicate that chronic subcutaneous administration of OEtA reduces bladder overactivity and improves urodynamic parameters supporting its application in research on pain anxiety and bladder dysfunction