Learn More
Cayman Chemical Oleoyl-L-carnitine (chloride), Purity: ≥95%, CAS: 31062-78-9, 25 mg
SDP
Catalog No. 502917305
Encompass_Preferred
A long-chain acylcarnitine and GlyT2 inhibitor (IC50 = 340 nM); selective for GlyT2 over GlyT1 (IC50 = >10,000 nM); plasma levels are increased in patients with chronic kidney disease or end-stage renal disease receiving incident hemodialysis and these are associated with cardiovascular mortality
Catalog No. 50-291-7305 Supplier Cayman Chemical Supplier No. 3901425 MG
A long-chain acylcarnitine and GlyT2 inhibitor (IC50 = 340 nM); selective for GlyT2 over GlyT1 (IC50 = >10,000 nM); plasma levels are increased in patients with chronic kidney disease or end-stage renal disease receiving incident hemodialysis and these are associated with cardiovascular mortality