Learn More
Cayman Chemical Oleoyl-L-carnItInchlorIde 5mg
SDP
Catalog No. 502918954
Encompass_Preferred
A long-chain acylcarnitine and GlyT2 inhibitor (IC50 = 340 nM); selective for GlyT2 over GlyT1 (IC50 = >10,000 nM); plasma levels are increased in patients with chronic kidney disease or end-stage renal disease receiving incident hemodialysis and these are associated with cardiovascular mortality
Catalog No. 50-291-8954 Supplier Cayman Chemical Supplier No. 390145 MG
A long-chain acylcarnitine and GlyT2 inhibitor (IC50 = 340 nM); selective for GlyT2 over GlyT1 (IC50 = >10,000 nM); plasma levels are increased in patients with chronic kidney disease or end-stage renal disease receiving incident hemodialysis and these are associated with cardiovascular mortality