Learn More
Cayman Chemical Oleoyl SerInol 50mg
SDP
Catalog No. 502955627
Encompass_Preferred
An analog of ceramide and a GPR119 agonist (EC50 = 12 µM); induces cell death in F-11 murine neuroblastoma and U-87 human astrocytoma cells at 100 µM; induces apoptosis in rapidly dividing cells and prevents teratoma formation in a neonatal mouse engraftment model of teratoma formation
Catalog No. 50-295-5627 Supplier Cayman Chemical Supplier No. 1363750 MG
An analog of ceramide and a GPR119 agonist (EC50 = 12 µM); induces cell death in F-11 murine neuroblastoma and U-87 human astrocytoma cells at 100 µM; induces apoptosis in rapidly dividing cells and prevents teratoma formation in a neonatal mouse engraftment model of teratoma formation