Learn More
Cayman Chemical Oleoyl Serinol, Purity: ≥98%, CAS: 72809-08-6, 5 mg
SDP
Catalog No. 502955212
Encompass_Preferred
A gut microbe-derived agonist of GPR119; stimulates GLP-1 secretion in vitro (IC50 = 12 µM); has a variety of meal-related effects in mice, including a reduction of plasma insulin, but not leptin, levels in ad libitum-fed mice and an increase in both plasma insulin and leptin levels in mice fasted for 12 hours, then refed; alters the circadian oscillations of mouse plasma phosphatidylcholine and ceramide levels in a molecular species-specific manner; reduces cell viability of cancer cells in vitro; induces formation of a complex including PAR-4 and PKCζ in embryoid body-derived cells, decreases the levels of Oct-4 and PAR-4
Catalog No. 50-295-5212 Supplier Cayman Chemical Supplier No. 136375 MG
A gut microbe-derived agonist of GPR119; stimulates GLP-1 secretion in vitro (IC50 = 12 µM); has a variety of meal-related effects in mice, including a reduction of plasma insulin, but not leptin, levels in ad libitum-fed mice and an increase in both plasma insulin and leptin levels in mice fasted for 12 hours, then refed; alters the circadian oscillations of mouse plasma phosphatidylcholine and ceramide levels in a molecular species-specific manner; reduces cell viability of cancer cells in vitro; induces formation of a complex including PAR-4 and PKCζ in embryoid body-derived cells, decreases the levels of Oct-4 and PAR-4