Learn More
Cayman Chemical Oleoyl Serotonin, Purity: ≥98%, CAS: 1002100-44-8, 5 mg
SDP
Catalog No. 502892509
Encompass_Preferred
A hybrid molecule patterned after N-arachidonoyl serotonin; inhibits capsaicin-induced TRPV1 channel activation (IC50 = 2.57 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM)
Catalog No. 50-289-2509 Supplier Cayman Chemical Supplier No. 90006295 MG
A hybrid molecule patterned after N-arachidonoyl serotonin; inhibits capsaicin-induced TRPV1 channel activation (IC50 = 2.57 μM) without blocking FAAH-mediated hydrolysis of arachidonoyl ethanolamine (IC50 > 50 μM)