Learn More
Cayman Chemical Oleoyl SerotonIn-d17 100UG
SDP
Catalog No. 502894606
Encompass_Preferred
An internal standard for the quantification of oleoyl serotonin by GC- or LC-MS
Catalog No. 50-289-4606 Supplier Cayman Chemical Supplier No. 9000694100 UG
An internal standard for the quantification of oleoyl serotonin by GC- or LC-MS