Learn More
Cayman Chemical Oleoyl Serotonin-d17, Purity: ≥99% deuterated forms (d1-d17), 1 mg
SDP
Catalog No. 502895191
Encompass_Preferred
An internal standard for the quantification of oleoyl serotonin by GC- or LC-MS
Catalog No. 50-289-5191 Supplier Cayman Chemical Supplier No. 90006941 MG
An internal standard for the quantification of oleoyl serotonin by GC- or LC-MS