Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oleoylcarnitine | 38677-66-6 | 99.95% | 425.64 | 1 MG
SDP

Catalog No. 5000436749
Encompass_Preferred

Oleoylcarnitine is a metabolite that accumulates when fatty acid β-oxidation is suppressed. It can enhance hepatocarcinogenesis by activating STAT3.

  • Enhances hepatocarcinogenesis via STAT3 activation.
  • Accumulates in obesity-driven HCC tissues and serum of high-fat diet-fed mice.
  • Increases in concentration in Dih10 cells with stable CPT2 knock down.
  • Contributes to hepatocarcinogenesis by conferring stem cell properties to cancer cells.
  • Purity: 99.95%.
  • Appearance: Solid, white to off-white.
  • Initial source: Endogenous metabolite.

Catalog No. 50-004-36749 Supplier Medchemexpress LLC Supplier No. HY1132611MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oleoylcarnitine is a metabolite that accumulates when fatty acid β-oxidation is suppressed. It can enhance hepatocarcinogenesis by activating STAT3.

  • Enhances hepatocarcinogenesis via STAT3 activation.
  • Accumulates in obesity-driven HCC tissues and serum of high-fat diet-fed mice.
  • Increases in concentration in Dih10 cells with stable CPT2 knock down.
  • Contributes to hepatocarcinogenesis by conferring stem cell properties to cancer cells.
  • Purity: 99.95%.
  • Appearance: Solid, white to off-white.
  • Initial source: Endogenous metabolite.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.