Learn More
Medchemexpress LLC HY-107542 10mg Medchemexpress, Oleoylethanolamide CAS:111-58-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10754210MG
Encompass_Preferred
Medchemexpress, HY-107542 10mg Oleoylethanolamide CAS:111-58-0 Oleoylethanolamide is a high affinity endogenous PPAR-α agonist, which plays an important role in the treatment of obesity and arteriosclerosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8432