Learn More
TARGETMOL CHEMICALS INC OLEOYLETHANOLAMIDE 25MG
SDP
Catalog No. 502581470
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oleoylethanolamide (N-Oleoylethanolamide) is a high affinity endogenous agonist of PPAR-(alpha). purity: 99%
Catalog No. 50-258-1470 Supplier TARGETMOL CHEMICALS INC Supplier No. T1229625MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Oleoylethanolamide (N-Oleoylethanolamide) is a high affinity endogenous agonist of PPAR-(alpha). purity: 99%