Learn More
AdipoGen Oleuropein
SDP
Supplier:  AdipoGen AGCN20538M025
Encompass_Preferred
Chemical. CAS 32619-42-4. Formula C25H32O13. MW 540.5. Oleuropein is a polyphenol from olive leaves and fruits that hydrolyzes to form hydroxytyrosol and tyrosol. Oleuropein exhibits antioxidant, anti-inflammatory, anticancer, antimicrobial, antifungal, antiviral, antiobesity, antidiabetic, antihypertensive, hypolipidemic, hepatoprotective and neuroprotective properties. Free radical-scavenging compound. Inhibits cell proliferation, cell growth, migration, invasion and angiogenesis and induces apoptosis and cell cycle arrest in cancer cells but not normal cells. Modulates gene expression and activity of a variety of different signaling proteins that play a role in proliferation and apoptosis. Induces autophagy by activating AMPK. Potential agent against metabolic syndrome. Reduces adipogenesis by antagonizing PPARgamma. Enhances UCP1 expression in brown adipose tissue in high-fat-diet-induced obese rats by activating beta-adrenergic signaling.
Catalog No. 50-256-2489