Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleyl Acetate 60.0+%, TCI America™
SDP

Catalog No. A093425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A093425ML 25 mL
A0934500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A093425ML Supplier TCI America Supplier No. A093425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 693-80-1
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.52
MDL Number MFCD00056326
InChI Key GYGAZRPDUOHMAF-KHPPLWFESA-N
Synonym Acetic Acid Oleyl Ester
PubChem CID 5363289
IUPAC Name (9Z)-octadec-9-en-1-yl acetate
SMILES CCCCCCCC\C=C/CCCCCCCCOC(C)=O

Specifications

Color Yellow
Boiling Point 186°C
Quantity 25 mL
Formula Weight 310.52
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Oleyl Acetate
EINECSNumber (2)-0762
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.