Learn More
Medchemexpress LLC Oleyl bromide | 6110-53-8 | 98.36% | 331.37 | 50 MG
SDP
Catalog No. 5000451898
Encompass_Preferred
Oleyl bromide ((Z)-1-Bromooctadec-9-ene) is a hydrophobic compound capable of undergoing a ring-opening reaction through a process known as "encapsulation." It can also serve as a starting material in the synthesis of oleylphosphonates.
  • Hydrophobic compound.
  • Can carry out ring-opening reactions via "encapsulation."
  • Used as a starting material in the synthesis of oleylphosphonates.
  • Ships at room temperature in continental US.
  • Pure form storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-51898 Supplier Medchemexpress LLC Supplier No. HYW01123850MG
Oleyl bromide ((Z)-1-Bromooctadec-9-ene) is a hydrophobic compound capable of undergoing a ring-opening reaction through a process known as "encapsulation." It can also serve as a starting material in the synthesis of oleylphosphonates.
  • Hydrophobic compound.
  • Can carry out ring-opening reactions via "encapsulation."
  • Used as a starting material in the synthesis of oleylphosphonates.
  • Ships at room temperature in continental US.
  • Pure form storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.