Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleyl Phosphate (Mono- and Di- Ester mixture), TCI America™
SDP

Catalog No. O014625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O014625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O014625ML Supplier TCI America Supplier No. O014625ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37310-83-1
MDL Number MFCD00135103
Synonym Phosphoric Acid Oleyl Ester

Specifications

Quantity 25 mL
Physical Form Liquid
Chemical Name or Material Oleyl Phosphate (Mono- and Di- Ester mixture)
EINECSNumber (2)-2014
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.