Learn More
Medchemexpress LLC Oleylamine | 112-90-3 | 90.0% | 267.49 | 100 G
SDP
Catalog No. 5000452866
Encompass_Preferred
Oleylamine is a versatile reagent employed for metal ion coordination and nanoparticle surface modification. It functions as a solvent, surfactant, and reducing agent in the synthesis of metal oxide nanoparticles. This compound is primarily utilized in nanomaterial synthesis, biomedical imaging, cancer cell targeting, and drug delivery research.
  • Regulates nanoparticle morphology, magnetization intensity, and water proton relaxation rate via thiol-ene "click" reaction.
  • Enhances the colloidal stability of nanoparticles in organic reagents.
  • Can exhibit concentration-dependent cytotoxicity towards human HeLa and MRC5 cell lines at certain concentrations.
Catalog No. 50-004-52866 Supplier Medchemexpress LLC Supplier No. HYW011696100G
Oleylamine is a versatile reagent employed for metal ion coordination and nanoparticle surface modification. It functions as a solvent, surfactant, and reducing agent in the synthesis of metal oxide nanoparticles. This compound is primarily utilized in nanomaterial synthesis, biomedical imaging, cancer cell targeting, and drug delivery research.
  • Regulates nanoparticle morphology, magnetization intensity, and water proton relaxation rate via thiol-ene "click" reaction.
  • Enhances the colloidal stability of nanoparticles in organic reagents.
  • Can exhibit concentration-dependent cytotoxicity towards human HeLa and MRC5 cell lines at certain concentrations.