Learn More
Enzo Life Sciences Oleyloxyethyl phosphorylcholine (10mg). CAS: 96720-06-8
SDP
Catalog No. 502010560
Encompass_Preferred
A potent and specific substrate analog PLA2 inhibitor (IC50=6.2 µM-porcine pancreatic PLA2). It inhibits human recombinant sPLA2 (IC50=13.7 µM) selectively over cPLA2. At 1 µM it completely blocked the myogenic response in perfused dog renal arcuate arteries. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-201-0560 Supplier Enzo Life Sciences Supplier No. BMLEI1180010
A potent and specific substrate analog PLA2 inhibitor (IC50=6.2 µM-porcine pancreatic PLA2). It inhibits human recombinant sPLA2 (IC50=13.7 µM) selectively over cPLA2. At 1 µM it completely blocked the myogenic response in perfused dog renal arcuate arteries. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol. Long Term Storage: -20°C.