Learn More
Aviva Systems Biology Corp OLFM4 Antibody - C-terminal region (ARP51203_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51203_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against OLFM4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human OLFM4. Alternate names: GC1, GW112, KIAA4294, OlfD, bA209J19.1, OLM4, hGC-1, hOLfD, UNQ362ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-53653