Learn More
Medchemexpress LLC OLFM4 Human HEK293 2ug
SDP
Catalog No. 5000196471
Encompass_Preferred
Recombinant human OLFM4 protein produced in HEK293 mammalian cells and supplied lyophilized with a C-terminal His tag. Designed for research use in biochemical and cell biology assays, the product is provided in microgram-scale sizes and formulated to preserve stability and activity when handled according to storage and reconstitution guidelines.
  • Expressed in HEK293 mammalian cells for human-like post-translational modifications.
  • C-terminal His tag facilitates detection and purification.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Purity greater than 80% as determined by reducing SDS-PAGE.
  • Recommended to reconstitute at ≥100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; aliquot and store at -80°C for extended storage.
Catalog No. 50-001-96471 Supplier Medchemexpress LLC Supplier No. HYP711792UG
Recombinant human OLFM4 protein produced in HEK293 mammalian cells and supplied lyophilized with a C-terminal His tag. Designed for research use in biochemical and cell biology assays, the product is provided in microgram-scale sizes and formulated to preserve stability and activity when handled according to storage and reconstitution guidelines.
  • Expressed in HEK293 mammalian cells for human-like post-translational modifications.
  • C-terminal His tag facilitates detection and purification.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Purity greater than 80% as determined by reducing SDS-PAGE.
  • Recommended to reconstitute at ≥100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; aliquot and store at -80°C for extended storage.